Water plus - total ATP, Clean-Trace™
Lieferant: 3M Food Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AQT200
MMMPAQT200EA 577 EUR InStock
MMMPAQT200
Water plus - total ATP, Clean-Trace™
Überwachungssysteme Hygiene-Überwachungssysteme
3M™ Clean-Trace™ Water Plus – Total ATP is designed to rapidly assess equipment sanitation processes and the efficiency of Clean In Place (CIP) procedures by the improved detection of organic contamination from microbial or product residues.
  • Flexible storage options: 12 months shelf life refrigerated (2 to 8 °C) or 2 months shelf life at room temperature (21 to 25 °C)
  • Rapid results allowing for corrective action to be taken immediately
  • Simple to use – ideal for busy production environments
  • Ability to track and trend results using 3M™ Clean-Trace™ Data Trending software
The test gives an objective indication of cleanliness in water samples in seconds. Designedfor use with the 3M™ Clean-Trace™ Luminometer, the new AQT200 formulation nowprovides the ability to gain more consistent and reliable results providing greater confidencein your hygiene monitoring procedures.The new and improved formulation also provides the flexibility to store AQT200 at your ownconvenience - room temperature for up to 2 months or refrigerated for up to 12 months.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge