Water positive control (30 ng), HWATP10, Clean-Trace™
Lieferant: 3M Food Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HWATP10
MMMPHWATP10EA 194 EUR InStock
MMMPHWATP10
Water positive control (30 ng), HWATP10, Clean-Trace™
Überwachungssysteme Hygiene-Überwachungssysteme
Designed to provide assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly.
  • Verifies the performance of reagents and instruments
  • Can be used as a quench test with 3M™ Clean-Trace™ Water ATP tests
Designed to enhance your hygiene monitoring procedures by providing assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly to ensure reliable results.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge