Molecular detection matrix control
Lieferant: 3M FOOD SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MDMC96NA
MMMPMDMC96NAEA 808 EUR InStock
MMMPMDMC96NA
Molecular detection matrix control
Tests Immunologische Tests
Kit contains 96 tests and is used with the 3M™ Molecular Detection System to check for inhibition of the matrix sample during validation periods, when adopting the 3M™ method or testing new or unknown matrices.
  • Verify that new or unknown matrices are not inhibitory to amplification
  • Ready-to-use and pre-dispensed reagents
  • Only two transfer steps after enrichment
  • Closed-tube system reduces risk of amplicon contamination
  • Designed to work with 3M™ proprietary enrichment broths to promote growth
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge