Swab-sampler with buffered peptone water broth
Lieferant: 3M Food Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RS96010BPW
MMMPRS96010BPWEA 181 EUR InStock
MMMPRS96010BPW
Swab-sampler with buffered peptone water broth
Tupfer Tupfer für die Probenahme
3M™ Swab-Sampler with 10 ml Buffered Peptone Water Broth is a convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Buffered Peptone Water Broth vial is ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Sample collection is easy with the attached swab and the vial provides ample headspace for sample addition and vortexing to release organisms from the swab. This sampling device is decontaminated through gamma irradiation for long shelf life. The 3M Swab-Sampler is made in the USA and is ISO 9001:2008 certified for quality assurance. It is intended for lab use only. The Hazard Analysis Critical Control Point (HACCP) approach is the most widely accepted food safety management protocol worldwide. Sample collection products from 3M are a crucial part of this approach to identify potential food-borne hazards and pinpoint where they reside. Portability, stable enrichment media, and easy-to-handle swabs and sponges are designed to test for a wide variety of potential hazards and deliver exceptionally accurate results. They are engineered to safely gather, hold, and secure contents from the collection site to the lab without leaking or compromising the sample.
Buffered Peptone Water Broth is used as a diluent or for non selective enrichment of microorganisms such as Salmonella species or E. coli from foods and environmental samples prior to selective enrichment and isolation. Microorganisms that may be present in a food product can be sub-lethally injured by food processing techniques and may not be recovered by direct selection techniques. Buffered Peptone Water Broth promotes the recovery of those microorganisms that may be injured.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge