Blood glucose lancets, VITREX® Soft
Lieferant: Vitrex Medical A/S
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
351213 352213 354213 357213
MODU351213EA 15 EUR InStock
MODU351213 MODU352213 MODU354213 MODU357213
Blood glucose lancets, VITREX® Soft
Blutlanzetten
Needle material is stainless steel grade 304/1 Cr18Ni9 and moulded body/cap material is LDPE (Low Density Polyethylene).
  • Good blood flow - Triple ground needle point
  • Sterilised by gamma irradiation at minimum 25kGy
  • Convenient boxes with dispensing feature
VITREX® Soft is designed for use with most popular types of finger lancing device.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge