Bi-metallic dial thermometer with Min/Max memory
Lieferant: MOINEAU
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4100T
MOIN4100TEA 48.9 EUR InStock
MOIN4100T
Bi-metallic dial thermometer with Min/Max memory
Thermometer Analoge Thermometer
Maximum/minimum dial thermometer, bi-metallic.
  • For wall mounting
  • Made from white ABS
Two markers show the maximum and minimum temperature until the next reset.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge