4040
5200
4045
4080
5220
5150
4090
4020
4021
4025
5130
4030
4047
4051
4055
4060
4061
4062
4100
4125
4130
4135
4150
5140
5160
5241
5251
5281
391-3880EA
51.4
EUR
InStock
391-3880
MOLL5200
MOLL4045
MOLL4080
MOLL5220
MOLL5150
MOLL4090
MOLL4020
MOLL4021
MOLL4025
MOLL5130
MOLL4030
MOLL4047
MOLL4051
MOLL4055
MOLL4060
MOLL4061
MOLL4062
MOLL4100
MOLL4125
MOLL4130
MOLL4135
MOLL4150
MOLL5140
MOLL5160
MOLL5241
MOLL5251
MOLL5281
Culture media, ready to use, in tubes
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
These sterile ready-to-use media are filled in autoclavable and break-proof glass tubes (20 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C they can be poured as agar plates in different sizes and thicknesses to meet individual demands.
- Delivered 50 ml in polycarbonate bottles or 20 ml in glass tubes