Culture media, ready to use, in tubes
Lieferant: MÍLLER SCHMELZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4040 5200 4045 4080 5220 5150 4090 4020 4021 4025 5130 4030 4047 4051 4055 4060 4061 4062 4100 4125 4130 4135 4150 5140 5160 5241 5251 5281
391-3880EA 51.4 EUR InStock
391-3880 MOLL5200 MOLL4045 MOLL4080 MOLL5220 MOLL5150 MOLL4090 MOLL4020 MOLL4021 MOLL4025 MOLL5130 MOLL4030 MOLL4047 MOLL4051 MOLL4055 MOLL4060 MOLL4061 MOLL4062 MOLL4100 MOLL4125 MOLL4130 MOLL4135 MOLL4150 MOLL5140 MOLL5160 MOLL5241 MOLL5251 MOLL5281
Culture media, ready to use, in tubes
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
These sterile ready-to-use media are filled in autoclavable and break-proof glass tubes (20 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C they can be poured as agar plates in different sizes and thicknesses to meet individual demands.
  • Delivered 50 ml in polycarbonate bottles or 20 ml in glass tubes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge