Safety dispensing canisters
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
16779 16780 16781
MSGA16779EA 394 EUR InStock
MSGA16779 MSGA16780 MSGA16781
Safety dispensing canisters
Sicherheitsbehälter
Canister, made of stainless steel, with tap and separate ventilation.
  • Manual pressure
  • Vacuum balancing
  • Self-closing tap
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge