Drum taps, stainless steel
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2795 2797
MSGA2795EA 217.5 EUR InStock
MSGA2795 MSGA2797
Drum taps, stainless steel
Armaturen und Spülbecken Armaturen
These drum taps are made of stainless steel.
  • Adjustable removal position
  • Self closing
  • Flame guard
  • PTFE seal
Bestellinformation: MSGA2797 includes a trailer fitting for decanting vessels.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge