Safety storage cabinets, Type 30, K-PHOENIX-30
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30343001
MSGA30343001EA 3433 EUR InStock
MSGA30343001
Safety storage cabinets, Type 30, K-PHOENIX-30
Möbel Lagerschränke
Safe and approved storage of aggressive and flammable hazardous substances in working areas.
  • Robust construction and longevity - triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Two in one - 2 compartments (vertical separation) allow the storage of flammable liquids and aggressive hazardous materials in one cabinet
  • User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use - doors lockable with profi le cylinder (integration in an existing locking system possible) / cylinder lock
Compartment for the storage of aggressive, non flammable hazardous materials, metal-free tray shelves and collecting sumps.
Bestellinformation: Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
Lieferumfang: Delivery prices available upon request.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge