Dispensing Can
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36488
MSGA36488EA 105 EUR InStock
MSGA36488
Dispensing Can
Sicherheitsbehälter
The dispensing can allows for safe, clean and comfortable dispensing of liquids in the workplace.
  • Resistant, chemically stable and UV-stable containers
  • 18 labels in 9 signal colours and a multilingual safety label for content labelling according to GHS
  • With stable handles for ergnomic handling
Includes 2 screw-on type outlets in different sizes.
Width: 173 mm
Height: 416 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge