Safety PE Storage Can with Dosing Tap
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40096
MSGA40096EA 92.5 EUR InStock
MSGA40096
Safety PE Storage Can with Dosing Tap
Sicherheitsbehälter
The PE Storage Can is ideal for the safe storage and internal transport of liquids, such as acids or lyes.
  • With fine dosing tap made of stainless steel (1.4301), includes vent, PTFE seal
  • Includes multilingual safety sticker to designate the contents
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge