Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

HTD Equilibrium Dialysis Device

Lieferant: Chemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

1103 1006 1104 1105
XXXX1103EA 0 EUR InStock
XXXX1103 IBSA1006 MSPP1104 XXXX1105
HTD Equilibrium Dialysis Device
Schläuche Dialyseschläuche
HTD reusable equilibrium dialysis device combines industry-leading accuracy and precision with an automation compatible design for a cost efficient, scalable solution.

  • Simultaneously process 1 to 96 assays, each with a 30 to 150 µl capacity to conserve valuable reagents
  • Compatible with a wide range of dialysis membrane pore sizes
  • Ready in minutes, achieves equilibrium in 4 to 6 hours on average
  • Automatable with standard robotic workstations and 96-well pipettors
  • Ready access to both the sample and dialysate sides of the membrane from the top of the device
  • Minimal non-specific binding due to 100% Teflon block construction
  • Reusable device is cost-effective, eco-friendly, and scalable
  • Compatible with a wide range of dialysis membrane pore sizes: 1 kDa, 3,5 kDa, 6 to 8 kDa, 10 kDa, 12 to 14 kDa, 25 kDa, 50 kDa

The HTD96B micro equilibrium device was developed at a Global Pharmaceutical Research laboratory to assess serum protein binding, microsomal protein binding, and tissue specific binding of drug candidates, conserving sample quantities, and achieving rapid equilibrium.

Reusable device combines industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solution.

Applications:
- Serum/plasma protein binding
- Microsome protein binding
- Relative binding of drug to specific tissues
- Receptor binding
- T3/T4 free fraction binding (hyperthyroidism) determinations

The 96C model has a shorter diffusion path with a 25 to 75 µl capacity and achieves equilibrium in 2 to 4 hours on average.

Additional specifications:
Dimensions: 170,1×116,8×38,1 mm
Weight: 0,54 kg

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge