2 vials BirA (20 ug ea) lyophilized **** 1 tube 500 ul BirA storage/reconstitution buffer 1 tube 5 mL of SuperMix buffer 1 tube 275 mg of ATP under argon gas 5 Empty Storage tubes 1 * 1 KIT
Lieferant: MARKET SOURCE PART PROCESS
2 vials BirA (20 ug ea) lyophilized **** 1 tube 500 ul BirA storage/reconstitution buffer 1 tube 5 mL of SuperMix buffer 1 tube 275 mg of ATP under argon gas 5 Empty Storage tubes 1 * 1 KIT
PRODUKT
Erweitert / Kompakt
2 vials BirA (20 ug ea) lyophilized **** 1 tube 500 ul BirA storage/reconstitution buffer 1 tube 5 mL of SuperMix buffer 1 tube 275 mg of ATP under argon gas 5 Empty Storage tubes 1 * 1 KIT MSPPBIRA500-RT 1 * 1 KIT
Hersteller-Artikelnummer:: BIRA500-RT
Lokale Artikelnummer:: MSPPBIRA500-RT
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder