qEV1 / 35 nm - 5 Pack With 1 mL sample loading, the new qEV1 / 35 nm uses Izons Gen 2 resin and has an optimal recover range of 35 nm to 350 nm 1 * 5 ST
Lieferant: MARKET SOURCE PART PROCESS
qEV1 / 35 nm - 5 Pack With 1 mL sample loading, the new qEV1 / 35 nm uses Izons Gen 2 resin and has an optimal recover range of 35 nm to 350 nm 1 * 5 ST
PRODUKT
Erweitert / Kompakt
qEV1 / 35 nm - 5 Pack With 1 mL sample loading, the new qEV1 / 35 nm uses Izons Gen 2 resin and has an optimal recover range of 35 nm to 350 nm 1 * 5 ST MSPPIC1-35 1 * 5 ST
Hersteller-Artikelnummer:: IC1-35
Lokale Artikelnummer:: MSPPIC1-35
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder