RheVeTec RheFLEX MB125 12.7-6.4 mm shrink ratio 2:1 heat shrink tubing 100% polyolefin, radiation crosslinked, colour blue Minimum shrink temperature 90C, operating temperature -55C to +125C 1 * 1 ST
Lieferant: MARKET SOURCE PART PROCESS
RheVeTec RheFLEX MB125 12.7-6.4 mm shrink ratio 2:1 heat shrink tubing 100% polyolefin, radiation crosslinked, colour blue Minimum shrink temperature 90C, operating temperature -55C to +125C 1 * 1 ST
PRODUKT
Erweitert / Kompakt
RheVeTec RheFLEX® MB125 12.7-6.4 mm shrink ratio 2:1 heat shrink tubing 100% polyolefin, radiation crosslinked, colour blue Minimum shrink temperature 90°C, operating temperature -55°C to +125°C 1 * 1 ST MSPPMB125-127-0004 1 * 1 ST
Hersteller-Artikelnummer:: MB125-127-0004
Lokale Artikelnummer:: MSPPMB125-127-0004
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder