VWR® Gold-N-Gel™, RNA dye
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N734-5MLEA 251 EUR InStock
N734-5ML
VWR® Gold-N-Gel™, RNA dye
Färbemittel Elektrophorese-Färbemittel
Gold-N-Gel RNA dye is compatible with standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illumination with UV light. A standard ethidium bromide filter can be used for gel documentation.
  • Non mutagenic, fluorescent in-gel stain for immediate visualisation of RNA bands on denaturing agarose gels
  • Immediate band visualisation with UV illumination - No post-run staining or destaining
  • Easy-to-use: Simply add to melted agarose prior to casting
Lieferumfang: Supplied as a 200X concentrate.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge