VWR®, Phenol-free total RNA purification kit
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N788-KITEA 809 EUR InStock
N788-KIT
VWR®, Phenol-free total RNA purification kit
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The phenol-free total RNA purification kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses.
  • Eliminate phenol and chloroform from RNA purification
  • Enhance recovery of total RNA, including microRNA species
  • Save time with a simple 20 minute protocol
The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The RNA is preferentially purified from other cellular components, such as proteins.
The purified RNA, of the highest integrity can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.
Lieferumfang: Contains lysis solution, 40 ml; wash solution, 22 ml; elution solution, 6 ml; 50× mini spin columns; and 50× collection tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..