Protective gas boxes for furnaces, N7 - N 641
Lieferant: NABERTHERM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
631000384
NABE631000384EA 0 EUR InStock
NABE631000384
Protective gas boxes for furnaces, N7 - N 641
Öfen Veraschungs- und Muffelöfen
Hardening boxes are suitable for heat treatment under protective gas and are equipped with a protective gas intlet and outlet. For furnace models such as N 61/H with downward opening doors, the gas ductway is laid through the upper section of the door collar and for larger furnaces with upward opening doors, the supply line is laid through the lower furnace collar.
  • Protective gas box with lid, protective gas inlet and outlet through the furnace collar and seal profile
  • Maximum temperature of 1100 °C
  • Charge thermocouple type K
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge