Charging Devices with and without Cooling Fan for N-Series
Lieferant: NABERTHERM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
631000528
NABE631000528EA 0 EUR InStock
NABE631000528
Charging Devices with and without Cooling Fan for N-Series
Öfen Hochtemperaturöfen
For charging larger workpieces and annealing boxes.
  • 4 casters, freely movable
  • Equipped with a rack at working height for temporary storage
  • Fixing lock for annealing bags (CWK)
  • CWK version with cooling fan (0.2 kW, 230 V)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge