Tanks, zylindrisch, mit Sperrhahn und Deckel, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11102-0005 11102-0007 11102-0010 11102-0015 11102-0030 11102-0055
NALG11102-0005EA 291 EUR InStock
NALG11102-0005 NALG11102-0007 NALG11102-0010 NALG11102-0015 NALG11102-0030 NALG11102-0055
Tanks, zylindrisch, mit Sperrhahn und Deckel, Nalgene®
Tanks Tanks, zylindrisch
These heavy-duty cylindrical LLDPE tanks with cover and spigot can withstand most acid, alcohols and bases. Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
  • Economical
  • Impact- and abrasion-resistant
  • Not autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge