Drehmomentschlüsselanschlüsse für Tefzel-Verschlüsse, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2195-2438
NALG2195-2438EA 363 EUR InStock
NALG2195-2438
Drehmomentschlüsselanschlüsse für Tefzel-Verschlüsse, Nalgene®
Schraubenschlüssel
Durable epoxy resin construction, specifically designed for Nalgene ETFE closures.
Proper torquing of Nalgene closures is critical to leakproof sealing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge