Kanister, flach, mit Hahn und Schraubverschluss, Lowboys™, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2324-0015
NALG2324-0015EA 465 EUR InStock
NALG2324-0015
Kanister, flach, mit Hahn und Schraubverschluss, Lowboys™, Nalgene®
Flaschen Kanister
Made of PP, translucent.
  • Can be stacked three high
  • Autoclave between uses to eliminate bacterial contamination
  • Convenient integral handgrip for safe, easy transport
Carboys have a low-profile design which fits easily into tight spaces, on shelves or in lab refrigerators. Ideal for storing and dispensing reagents.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge