Spritzflaschen, weithalsig, farblich codiert, Unitary™, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2423-0500
NALG2423-0500EA 58.1 EUR InStock
NALG2423-0500
Spritzflaschen, weithalsig, farblich codiert, Unitary™, Nalgene®
Flaschen Spritzflaschen
LDPE, translucent, with PP closure.
  • Good chemical resistance
  • Good temperature resistance
Coded Unitary LDPE wash bottles, which come in a convenient, pre-packaged and easy-to-identify assortment. Red, white, blue or yellow closures/stems on these flexible unitary bottles permit easy content identification.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge