Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bottles, wide neck, HDPE, round, with screw cap, IP2

Lieferant: Thermo Fisher Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

312199-0004 312199-0008 312199-0016 312199-0032
NALG312199-0004EA 1830 EUR InStock
NALG312199-0004 NALG312199-0008 NALG312199-0016 NALG312199-0032
Bottles, wide neck, HDPE, round, with screw cap, IP2
Flaschen Kunststoffflaschen
Wide-mouth HDPE IP 2 bottles, translucent with PP screw cap.

  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof when used with Nalgene closures
  • Round shape

Safe, durable, economical bottles for the transport and packaging of hazardous goods. Recommended for customers who wish to put together and certify their own packaging system. Extra-wide mouth for simple filling and emptying of liquids and dry substances. The sturdy walls withstand cracking and damage from impact caused by shocks during transport. The bottom of the bottle incorporates IP2 marking, material, volume and registration notch. Evaluated at 15 psi per 49 CFR 173.27 © (2), ICAO Technical Instructions Part 4; 1.1.6 and IATA Dangerous Goods Regulations, Section 5.0.2.9.

Verpackung: Bottles and closures are bulk packed in separate polybags within the same carton.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge