Tamper evident closures for PET/PETG square media bottles, HDPE
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
342141-0384
NALG342141-0384EA 206 EUR InStock
NALG342141-0384
Tamper evident closures for PET/PETG square media bottles, HDPE
Verschlüsse Flaschenverschlüsse
Closure, tamper evident, specially designed for use with PET/PETG square media bottles.
  • Natural HDPE closures are ADCF (Animal Derived Component-Free)
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish and leakproof guarantee
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 10⁻⁶ Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labelling
Verpackung: Closures are sterile and bulk packed.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge