Bottles, narrow neck, round, HDPE, Boston type, with thread, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362002-0002 362002-0004 362002-0001 362002-0008 362002-0016
NALG362002-0002EA 974 EUR InStock
NALG362002-0002 NALG362002-0004 NALG362002-0001 NALG362002-0008 NALG362002-0016
Bottles, narrow neck, round, HDPE, Boston type, with thread, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Ideal for storing, packaging and transporting liquids. Robust, strong and stable.
Zertifizierungen: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge