Diagnostikflaschen, enghalsig, rund, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362002-9050 362002-9125 362002-9025
NALG362002-9050EA 1173 EUR InStock
NALG362002-9050 NALG362002-9125 NALG362002-9025
Diagnostikflaschen, enghalsig, rund, Nalgene®
Flaschen Kunststoffflaschen
Bottles without closure, used to store and ship small volumes of valuable reagents.
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof when assembled with Nalgene closure
Natural, translucent high-density polyethylene, durable, uniform walls resistant to splitting or puncturing, A sample product certificate providing cytotoxicity, USP Class VI, FDA compliance 21 CFR and other regulatory compliance information is available or can be requested from our certificates of compliance database.
Zertifizierungen: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP class VI.
Bestellinformation: Coloured closures are ordered seperately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge