Diagnostikflaschen, enghalsig, rund, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362004-9125 362004-9050 362004-9025
NALG362004-9125EA 1640 EUR InStock
NALG362004-9125 NALG362004-9050 NALG362004-9025
Diagnostikflaschen, enghalsig, rund, Nalgene®
Flaschen Kunststoffflaschen
Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ opaque amber HDPE diagnostic bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
  • Durable, non-sterile and leakproof
  • Non-pyrogenic and non-cytotoxic
  • Uniform walls are resistant to splitting or puncturing
Bottles provide light resistance and used in test kits and many small-volume applications.
Zertifizierungen: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Bestellinformation: Product certificate providing cytotoxicity, USP Class VI, FDA Compliance - 21 CFR and other regulatory compliance information is provided based upon request.
Verpackung: Bottles are bulk packed without closures for 2000 per case. Coloured closures should be order seperately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge