Diagnostikflaschen, enghalsig, rund, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362008-9125 362008-9025 362008-9050
NALG362008-9125EA 1720 EUR InStock
NALG362008-9125 NALG362008-9025 NALG362008-9050
Diagnostikflaschen, enghalsig, rund, Nalgene®
Flaschen Kunststoffflaschen
Opaque bottles without closure, used to store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples.
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof when assembled with Nalgene closure
Zertifizierungen: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
Bestellinformation: Coloured closures are ordered seperately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge