Biotainer® Replacement Screw Caps for Bottles, InVitro™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362515-0480
NALG362515-0480EA 3460 EUR InStock
NALG362515-0480
Biotainer® Replacement Screw Caps for Bottles, InVitro™
Verschlüsse Flaschenverschlüsse
Protect the contents in your Thermo Scientific™ Biotainer™ bottles and carboys. Thermo Scientific™ Nalgene™ sterile polypropylene biotainer replacement closures are for 1, 2, 5, 10 and 20 L polycarbonate biotainers.
  • Designed to fit only nalgene bottles to provide a guaranteed leakproof system
  • Polypropylene with silicone liner
  • Closure size: 48 mm
  • Packaged individually: 1 per bag, 6 bags or 50 per case, 300 in a case
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge