Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Non-phthalate PVC vacuum tubing, Nalgene™

Lieferant: Thermo Fisher Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

8702-0055 8702-0065 8702-1065 8702-0145 8702-1145 8702-0210 8702-1210
NALG8702-0055EA 239 EUR InStock
NALG8702-0055 NALG8702-0065 NALG8702-1065 NALG8702-0145 NALG8702-1145 NALG8702-0210 NALG8702-1210
Non-phthalate PVC vacuum tubing, Nalgene™
Schläuche Vakuum-Schläuche und Druck-Schläuche
Nalgene™ lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.

  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
  • Thick-walled PVC tubing withstands a full vacuum of 760 mm Hg at room temperature

Nalgene™ Non-phthalate PVC vacuum tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening and discolouration from a wide range of laboratory chemicals (demonstrates dimensional stability) and does not impart odour or taste.

Zertifizierungen: Meets food additives amendment of the federal food, drug, and cosmetic act. Meets the requirements of the U.S. pharmacopoeia class VI biological tests for plastics, USP <88>. Complies with FDA CFR21 for food packaging, USP VI, cytotoxicity, REACH, latex-free, RoHS, and California proposition 65 standards.

Achtung! Bitte beachten: Not intended for medical use.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge