Establish a leak-tight fit with metric fittings. Nalgene lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
- Compare to Tygon® E-3603 tubing
- Marked with Int.Ø. and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
- Durometer hardness is shore A, 60
- Offered in true metric sizes to fit your lab instrumentation and equipment
Nalgene™ Metric non-phthalate PVC tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening and discolouration from a wide range of laboratory chemicals (demonstrates dimensional stability) and does not impart odour or taste making this tubing ideal for a wide range of non-medial, general lab, research and pharmaceutical applications.
Zertifizierungen: Meets food additives amendment of the federal food, drug, and cosmetic act. Meets the requirements of the U.S. pharmacopoeia class VI biological tests for plastics, USP <88>. Complies with FDA CFR21 for food packaging, USP VI, cytotoxicity, REACH, latex-free, RoHS, and california proposition 65 standards
Achtung! Bitte beachten: Not intended for medical use.