Stopcocks, needle, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96423-0100
NALG96423-0100EA 253 EUR InStock
NALG96423-0100
Stopcocks, needle, Nalgene®
Hähne
Spigot is installed on a threaded HDPE boss which is welded onto the tank at the factory.
  • Boss fits only Nalgener tanks
  • Spigot Material: Polypropylene
  • Accepts ⁵/₈" inner Ø tubing
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge