Mini spore discs
Lieferant: CROSSTEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DN18-06
NAMSDN18-06EA 510 EUR InStock
NAMSDN18-06
Mini spore discs
Sterilisationsindikatoren
Inoculated carrier mini spore discs for monitoring EO and dry heat processes.
  • An inoculated carrier, 3 mm disc, of Bacillus atrophaeus (cell Line 9372)
  • Primary packaging in bulk
The mini spore discs are designed to be placed directly into a device and utilised to monitor EO and dry heat sterilisation process efficacy.
The mini spore discs are labelled for industrial use only.
Zertifizierungen: ISO 11138-1 Sterilisation of health care products – Biological indicators – Part 1: General requirements.
ISO 11138-2 Sterilisation of health care products – Biological indicators – Part 2: Biological indicators for ethylene oxide sterilisation processes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge