Self contained biological indicators (SCBI)
Lieferant: CROSSTEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SCS-06 SCS-05
NAMSSCS-06EA 677 EUR InStock
NAMSSCS-06 NAMSSCS-05
Self contained biological indicators (SCBI)
Sterilisationsindikatoren
Self-contained biological indicators (SCBIs) for monitoring steam sterilisation consist of a durable PC vial with cap, a crushable glass ampoule with Trypticase® soy broth (TSB) modified with pH indicator, and a disc inoculated with Geobacillus stearothermophilus spores.
  • Reduced incubation time of 24 hours
  • Visually distinct positive results indicated by the media transitioning from purple to yellow
  • Chemical indicator on label provides instant indication of exposure
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Zertifizierungen: Complies to ISO 11138-1, ISO 11138-3, USP.
Achtung! Bitte beachten: For industrial use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge