Sludge nabber
Lieferant: NASCO FORT ATKINSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B01229WA
NASCB01229WAEA 255 EUR InStock
NASCB01229WA
Sludge nabber
Probenehmer Probenehmer für Flüssigkeiten
Sample bottles of various sizes are held via an adjustable locking device.
  • Makes sample collection of liquids fast and easy
  • Flexible with adjustable aluminum pole
  • PU holder with a plastic snapper ring
Suitable for numerous requirements including chlorine in solution – Can also be used for BOD.
Lieferumfang: Supplied with a 500 ml PE bottle.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge