Volumetric flasks, class A, blue markings
Lieferant: NEUBERT VOLUME GLASSWAERE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0293-04-0400 0293-04-0015 0293-04-0010 0293-04-0005 0293-04-0901 0293-04-0150 0293-04-0601 0293-04-035014
NEUB0293-04-0400EA 52.6 EUR InStock
NEUB0293-04-0400 NEUB0293-04-0015 NEUB0293-04-0010 NEUB0293-04-0005 NEUB0293-04-0901 NEUB0293-04-0150 NEUB0293-04-0601 NEUB0293-04-035014
Volumetric flasks, class A, blue markings
Kolben Messkolben
Borosilicate glass, grade 3.3, with PE stopper.
  • With one mark
  • Good chemical resistance
  • Good temperature resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge