Sublimation apparatus
Lieferant: NEUBERT VOLUME GLASSWAERE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2485-06-29
NEUB2485-06-29EA 265 EUR InStock
NEUB2485-06-29
Sublimation apparatus
Destillationsbauteile Destillations-Bausätze
This sublimation apparatus, made of borosilicate glass 3.3, is for working under vacuum.Total height: 200 mm.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge