Refill spill kits, in extra-large response cart, HazMat
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KITR304
NEWPKITR304EA 1251.25 EUR InStock
NEWPKITR304
Refill spill kits, in extra-large response cart, HazMat
Absorptionsmaterial
The refills for PIG® HazMat contain PIG HazMat absorbents which are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulphuric acid and 30% sodium hydroxide.
  • PIG® HazMat socks stop spreading spills; PIG® HazMat pads and pillows absorb; PIG® HazMat pulp cleans up residue on rough surfaces
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
  • Four easy-access compartments hold supplies to handle large spills
Kit includes: 2× dikes (130 mm×3 m), 7× socks (80×3 m), 36× socks (80×1170 mm), 100× heavy weight mat pads (380×510 mm), 15× pillows (410×430 mm), 2×2,9 kg pulp, 30× PE disposable bags (910×1520 mm), 6× tamperproof seals (150×55 mm).
Verpackung: Packed in sturdy, tri-wall cardboard.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge