PIG® Sticky Steps® mat
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MAT566-BL
NEWPMAT566-BLEA 110 EUR InStock
NEWPMAT566-BL
PIG® Sticky Steps® mat
Absorptionsmaterial
Multi-sheet floor mat grabs grime from shoes and wheels to keep dirt out of offices, cleanrooms and other areas where clean floors are vital.
  • High-tack disposable mat pulls dirt and debris from shoes and wheels to prevent tracking into offices
  • Used sheets tear off easily and individually
  • Numbered pull-tab system shows when you're running low
  • Low-profile pack adheres to floor with easy-to-use backing
  • Each 4 mil polyethylene sheet is nonallergenic, nonodourous and nondrying
  • Walking surface: Sticky top
Weight: 6,35 kg
Thickness: 100 micron
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge