Roll top hardcover spill pallet, PIG®
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PAK601-AWO PAK601-BWG-AWO
NEWPPAK601-AWOEA 1546.5 EUR InStock
NEWPPAK601-AWO NEWPPAK601-BWG-AWO
Roll top hardcover spill pallet, PIG®
Fasslager
These roll top hardcovers can be used for all-weather protection and liquid containment. Durable swing-out doors on front offers easy loading and unloading.
  • Weather-resistant design prevents contaminants from entering stormwater
  • Heavy-duty construction allows hardcover and pallet to be lifted by a forklift with drums in place
  • Low-density polyethylene (LDPE) with UV inhibitors resists UV rays, rust, corrosion and most chemicals for long, trouble-free life
  • Doors and roll top are available with a padlock
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge