Spill containment pools, PIG®
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PAK925 PAK926 PAK927 PAK928
NEWPPAK925EA 125 EUR InStock
NEWPPAK925 NEWPPAK926 NEWPPAK927 NEWPPAK928
Spill containment pools, PIG®
Absorptionsmaterial
These containment pools are ideal for quick deployment to catch the contents of blown hydraulic lines, punctured tanks, leaking containers and decontamination operations. To store our pop-up containment pool in the vehicle to be prepared for spill emergencies on the road.
  • Made of heavy-duty, 18 oz. vinyl-coated polyester provides high resistance to tears and punctures
  • Rim has foam ring that rises with level of liquid contained
  • Deploys instantly for use in an emergency; decontaminate for reuse
  • Pliable material folds easily for storage on a truck or emergency response vehicle
  • Excellent way to equip the fleet for punctured fuel tanks or drums while on the road
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge