PIG® Drive-Over DrainBlocker® Drain Covers
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PLR901 PLR906
NEWPPLR901EA 307.5 EUR InStock
NEWPPLR901 NEWPPLR906
PIG® Drive-Over DrainBlocker® Drain Covers
Absorptionsmaterial
PIG® DrainBlocker® Covers deploy quickly and create a tight, temporary seal over drains to keep out unwanted liquids. Ideal for heavy outdoor vehicle traffic.
  • Trust this drive-over drain cover - the proven-toughest on the market - to resist everything from punctures, stretching and tearing to prolonged exposure to weather, UV/IR light and oils like diesel fuel and gasoline, plus many chemicals
  • Maintain a professional look for longer since this drive-over drain cover is designed to wipe clean and get back on the job
  • Use it how you need it, either leaving it down as a preventive measure in leak- and spill-prone areas or storing it for emergency spill response
  • Respond faster to spills by conveniently carrying the cover in the included nylon storage bag
Confidently block drains from spills and water with this outdoor drain cover's super-sealing bottom layer that conforms perfectly for a tight seal while the HogHyde® rubber top layer withstands years and years of repeated heavy vehicle traffic.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge