Cinchable leak diverter, PIG®
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TLS182
NEWPTLS182EA 60.5 EUR InStock
NEWPTLS182
Cinchable leak diverter, PIG®
Absorptionsmaterial
The leak diverter made from PVC/polyester features drawstrings which cinch tightly around valves, fittings and other oddly shaped components to capture leaks at the source.
  • Compact size for leaks in tight spaces
  • Non-permeable material is vinyl-coated for strength and durability
  • Resists punctures and mildew for long life
  • UV resistant up to 12 months for long-term outdoor use without degradation
Accepts a standard 3/4" hose connection to safely channel liquid to floor drain or collection container (hose sold separately). Flow rate of 12,6 litres per minute when used with a 1/2" ID hose and can be used with 1,9 to 10 cm pipes.
Lieferumfang: Includes 1× leak diverter and 1× 3/4" drain fitting.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge