Cleanroom shoe cover, PP/CPE, BioClean Dual™
Lieferant: Nitritex
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S-BDOS
NITRS-BDOSEA 443 EUR InStock
NITRS-BDOS
Cleanroom shoe cover, PP/CPE, BioClean Dual™
Einweg-Kleidung
Disposable shoe covers made from BioClean Dual™, a low linting spunbonded non-woven polypropylene fabric with embossed cast polyethylene sole for a secure footing.
  • Durable
  • Heavy duty slip-resistant sole
  • Latex-free elastic
Suitable for use in ISO Class 4 environment.
Zertifizierungen: Manufactured in a facility holding ISO 9001:2008 certification and processed in an NEBB certified ISO Class 4 environment.
Verpackung: One pair per sealed inner PE bag, 15 inner bags per sealed outer bag, 10 outer bags per lined carton (150 pairs).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge