Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-p97/VCP Mouse Monoclonal Antibody [clone: Hs-14]

Lieferant: Novus Biologicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

NB500-455
NOVUNB500-455EA 456 EUR InStock
NOVUNB500-455
Anti-p97/VCP Mouse Monoclonal Antibody [clone: Hs-14]
Antikörper
The p97/VCP Antibody (Hs-14) from Novus Biologicals is a mouse monoclonal antibody to p97/VCP. This antibody reacts with human, mouse. The p97/VCP Antibody (Hs-14) has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Type: Primary
Antigen: p97/VCP
Clonality: Monoclonal
Clone: Hs-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge