Anti-AU5 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NB600-458
NOVUNB600-458EA 594 EUR InStock
NOVUNB600-458
Anti-AU5 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal AU5 Epitope Tag Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Non-species specific.
Type: Primary
Antigen: AU5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: TDFYLK
Host: Rabbit
Isotype:
Reactivity: pan
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge