Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD155/PVR Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

NBP1-88131-25
NOVUNBP1-88131-25EA 443 EUR InStock
NOVUNBP1-88131-25
Anti-CD155/PVR Rabbit Polyclonal Antibody
Antikörper
The CD155/PVR Antibody from Novus Biologicals is a rabbit polyclonal antibody to CD155/PVR. This antibody reacts with human. The CD155/PVR Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Polio Virus Receptor (PVR) is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or alpha and delta) encode cell surface molecules of about 43 kDa and 45 kDa polypeptide backbone, respectively. Two other splice variants, beta and gamma encode secreted molecules of about 44 kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues.

Achtung! Bitte beachten: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: CD155/PVR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge